Assay Details
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257293.2 | XP_006257355.1 | 6 - 7 | 884 | 79 | |
GenBank mRNA | FQ212519.1 | - | 2 - 3 | 211 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
