Assay Details
Gene Symbol: Slc13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031746.1 | NP_113934.1 | 6 - 7 | 926 | 62 | |
XM_006246947.2 | XP_006247009.1 | 6 - 7 | 918 | 62 | ||
GenBank mRNA | AB001321.1 | - | 6 - 7 | 929 | 62 | |
AF058714.1 | - | 6 - 7 | 926 | 62 | ||
U51153.1 | - | 6 - 7 | 916 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
