Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195505.1 | NP_001182434.1 | 563 | 115 | ||
NM_001195506.1 | NP_001182435.1 | 434 | 115 | |||
NM_019257.3 | NP_062130.2 | 438 | 115 | |||
XM_008764878.1 | XP_008763100.1 | 4 - 5 | 614 | 115 | ||
XM_008764879.1 | XP_008763101.1 | 4 - 5 | 485 | 115 | ||
GenBank mRNA | AF020683.1 | - | 4 - 5 | 500 | 115 | |
BC058479.1 | - | 432 | 115 | |||
FM049719.1 | - | 4 - 5 | 526 | 115 | ||
FM059586.1 | - | 4 - 5 | 412 | 115 | ||
FM081441.1 | - | 4 - 5 | 438 | 115 | ||
L13635.1 | - | 534 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
