Assay Details
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107049.1 | NP_001100519.1 | 8 - 9 | 1263 | 66 | |
XM_006247310.2 | XP_006247372.1 | 8 - 9 | 2117 | 66 | ||
GenBank mRNA | BC162060.1 | - | 8 - 9 | 1206 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
