Assay Details
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012832.3 | NP_036964.3 | 3 - 4 | 242 | 90 | |
XM_008759498.1 | XP_008757720.1 | 3 - 4 | 302 | 90 | ||
GenBank mRNA | AY574256.1 | - | 3 - 4 | 242 | 90 | |
AY671974.1 | - | 3 - 4 | 245 | 90 | ||
L31619.1 | - | 3 - 4 | 264 | 90 | ||
S53987.1 | - | 283 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
