Assay Details
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177421.3 | NP_803156.2 | 2 - 3 | 726 | 55 | |
XM_006251965.1 | XP_006252027.1 | 2 - 3 | 1364 | 55 | ||
GenBank mRNA | AB040056.1 | - | 2 - 3 | 733 | 55 | |
BC099072.1 | - | 2 - 3 | 726 | 55 | ||
FQ212273.1 | - | 2 - 3 | 728 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
