Assay Details
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031237.1 | NP_112516.1 | 1 - 2 | 292 | 67 | |
XM_006233333.1 | XP_006233395.1 | 2 - 3 | 284 | 67 | ||
XM_006233334.1 | XP_006233396.1 | 2 - 3 | 431 | 67 | ||
XM_006233335.1 | XP_006233397.1 | 2 - 3 | 238 | 67 | ||
XM_006233336.2 | XP_006233398.1 | 2 - 3 | 284 | 67 | ||
XM_006233337.1 | XP_006233399.1 | 2 - 3 | 431 | 67 | ||
XM_006233338.1 | XP_006233400.1 | 2 - 3 | 269 | 67 | ||
GenBank mRNA | AB006852.1 | - | 1 - 2 | 292 | 67 | |
AY724500.1 | - | 2 - 3 | 239 | 67 | ||
BC072696.1 | - | 2 - 3 | 215 | 67 | ||
FQ233355.1 | - | 2 - 3 | 259 | 67 | ||
U13175.1 | - | 1 - 2 | 146 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB006852.1, BC072696.1, NM_031237.1, U13175.1, XM_006233333.1, XM_006233334.1, XM_006233335.1, XM_006233336.2, XM_006233337.1, XM_006233338.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
