Assay Details
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031616.1 | NP_113804.1 | 3 - 4 | 239 | 158 | |
GenBank mRNA | AF013965.1 | - | 3 - 4 | 479 | 158 | |
AF013967.1 | - | 3 - 4 | 239 | 158 | ||
BC087012.1 | - | 3 - 4 | 225 | 158 | ||
FQ211774.1 | - | 3 - 4 | 265 | 158 | ||
FQ211917.1 | - | 3 - 4 | 265 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
