Assay Details
Gene Symbol: Klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228377.2 | XP_006228439.1 | 4 - 5 | 743 | 87 | |
XM_006228378.2 | XP_006228440.1 | 4 - 5 | 743 | 87 | ||
XM_006228379.2 | XP_006228441.1 | 4 - 5 | 665 | 87 | ||
XM_006228380.2 | XP_006228442.1 | 4 - 5 | 665 | 87 | ||
GenBank mRNA | BC078736.1 | - | 4 - 5 | 603 | 87 | |
BC097284.1 | - | 4 - 5 | 595 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
