Assay Details
Gene Symbol: Gtf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031041.1 | NP_112303.1 | 6 - 7 | 831 | 63 | |
GenBank mRNA | BC085345.1 | - | 6 - 7 | 836 | 63 | |
FQ229769.1 | - | 6 - 7 | 820 | 63 | ||
X65948.1 | - | 6 - 7 | 831 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
