Assay Details
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025689.1 | NP_001020860.1 | 6 - 7 | 592 | 81 | |
GenBank mRNA | BC097427.1 | - | 6 - 7 | 592 | 81 | |
BC158645.1 | - | 6 - 7 | 540 | 81 | ||
FQ228714.1 | - | 6 - 7 | 568 | 81 | ||
FQ228827.1 | - | 6 - 7 | 568 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
