Assay Details
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108272.2 | NP_001101742.1 | 1 - 2 | 91 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108272.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
