Assay Details
Gene Symbol: Nat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108466.1 | NP_001101936.1 | 1 - 2 | 436 | 82 | |
XM_006228268.2 | XP_006228330.1 | 2 - 3 | 386 | 82 | ||
XM_006228269.2 | XP_006228331.1 | 2 - 3 | 383 | 82 | ||
XM_006228270.1 | XP_006228332.1 | 2 - 3 | 337 | 82 | ||
XM_006228271.1 | XP_006228333.1 | 2 - 3 | 334 | 82 | ||
XM_006228272.2 | XP_006228334.1 | 2 - 3 | 690 | 82 | ||
XM_006228273.2 | XP_006228335.1 | 2 - 3 | 687 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
