Assay Details
Gene Symbol: Emc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234705.1 | XP_006234767.1 | 1 - 2 | 309 | 73 | |
GenBank mRNA | FQ212852.1 | - | 1 - 2 | 276 | 73 | |
FQ214203.1 | - | 1 - 2 | 276 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
