Assay Details
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276304.1 | NP_001263233.1 | 10 - 11 | 2820 | 57 | |
XM_008763989.1 | XP_008762211.1 | 11 - 12 | 4155 | 57 | ||
GenBank mRNA | AY530298.1 | - | 9 - 10 | 2716 | 57 | |
AY530299.1 | - | 9 - 10 | 2683 | 57 | ||
U67139.1 | - | 10 - 11 | 2820 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P072054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16123 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P072054 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P072054 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P072054 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P840999 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73281 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95728 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102936 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701746912 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10872143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872140 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872152 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10872139 | - | - |
Rat Genome 230 2.0 Array | 1376233_at | - | - |
Rat Genome 230 2.0 Array | 1370533_at | - | - |
Rat Genome 230 2.0 Array | 1370533_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376233_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376233_at | - | - |
Affymetrix HT Rat Focus Array | 1370533_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370533_at | - | - |
Affymetrix HT Rat Focus Array | 1376233_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500010585.rn.1 | TC0500001493.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500010583.rn.1 | TC0500001493.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500010584.rn.1 | TC0500001493.rn.1 | RTA TC |
Rat Expression Set 230 | 1376233_PM_at | - | - |
Rat Expression Set 230 | 1370533_at | - | - |
Rat Expression Set 230 | 1370533_PM_at | - | - |
Rat Expression Set 230 | 1376233_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10872139 | - | - |
Rat Genome U34 Set | U67139_at | - | - |
Rat Genome U34 Set | rc_AA942850_s_at | - | - |
Rat Genome U34 Set | rc_AI227989_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21160335 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369126 | - | - |
RatRef-12 Expression BeadChip | ILMN_53184 | - | - |
