Assay Details
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106131.1 | NP_001099601.1 | 10 - 11 | 2564 | 80 | |
XM_006254942.2 | XP_006255004.1 | 10 - 11 | 3669 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
