Assay Details
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106131.1 | NP_001099601.1 | 9 - 10 | 2354 | 79 | |
XM_006254942.2 | XP_006255004.1 | 9 - 10 | 3459 | 79 | ||
XM_006254943.2 | XP_006255005.1 | 9 - 10 | 3460 | 79 | ||
XM_006254944.2 | XP_006255006.1 | 9 - 10 | 3461 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
