Assay Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106924.1 | NP_001100394.1 | 5 - 6 | 918 | 116 | |
XM_006245373.2 | XP_006245435.1 | 6 - 7 | 1067 | 116 | ||
XM_006245374.2 | XP_006245436.1 | 6 - 7 | 1210 | 116 | ||
XM_006245375.2 | XP_006245437.1 | 6 - 7 | 969 | 116 | ||
GenBank mRNA | FQ225617.1 | - | 2 - 3 | 234 | 116 | |
FQ226825.1 | - | 1 - 2 | 60 | 116 | ||
FQ232683.1 | - | 2 - 3 | 234 | 116 | ||
FQ233663.1 | - | 1 - 2 | 59 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
