Assay Details
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014219.1 | NP_001014241.1 | 2 - 3 | 554 | 68 | |
XM_006245248.2 | XP_006245310.1 | 2 - 3 | 607 | 68 | ||
GenBank mRNA | BC081973.1 | - | 2 - 3 | 554 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
