Assay Details
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108808.1 | NP_001102278.1 | 4 - 5 | 943 | 121 | |
GenBank mRNA | BC158817.1 | - | 550 | 121 | ||
FQ219916.1 | - | 623 | 121 | |||
FQ220431.1 | - | 666 | 121 | |||
FQ231706.1 | - | 635 | 121 | |||
FQ232529.1 | - | 635 | 121 | |||
FQ233479.1 | - | 571 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
