Assay Details
Gene Symbol: Myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077656.1 | NP_001071124.1 | 1 - 2 | 243 | 87 | |
GenBank mRNA | CB809837.1 | - | 1 - 2 | 316 | 87 | |
FQ214580.1 | - | 1 - 2 | 370 | 87 | ||
FQ214604.1 | - | 1 - 2 | 288 | 87 | ||
FQ214608.1 | - | 1 - 2 | 248 | 87 | ||
FQ214631.1 | - | 1 - 2 | 230 | 87 | ||
FQ214712.1 | - | 1 - 2 | 251 | 87 | ||
FQ214723.1 | - | 1 - 2 | 251 | 87 | ||
FQ214727.1 | - | 1 - 2 | 251 | 87 | ||
FQ214742.1 | - | 1 - 2 | 251 | 87 | ||
FQ214939.1 | - | 1 - 2 | 248 | 87 | ||
FQ215050.1 | - | 1 - 2 | 251 | 87 | ||
FQ215083.1 | - | 1 - 2 | 248 | 87 | ||
FQ215201.1 | - | 1 - 2 | 248 | 87 | ||
FQ215435.1 | - | 1 - 2 | 248 | 87 | ||
FQ215563.1 | - | 1 - 2 | 248 | 87 | ||
FQ215610.1 | - | 1 - 2 | 251 | 87 | ||
FQ215774.1 | - | 1 - 2 | 248 | 87 | ||
FQ215848.1 | - | 1 - 2 | 248 | 87 | ||
FQ215875.1 | - | 1 - 2 | 248 | 87 | ||
FQ215933.1 | - | 1 - 2 | 251 | 87 | ||
FQ216034.1 | - | 1 - 2 | 251 | 87 | ||
FQ216109.1 | - | 1 - 2 | 248 | 87 | ||
FQ216181.1 | - | 1 - 2 | 230 | 87 | ||
FQ216218.1 | - | 1 - 2 | 248 | 87 | ||
FQ216229.1 | - | 1 - 2 | 518 | 87 | ||
FQ216407.1 | - | 1 - 2 | 419 | 87 | ||
FQ216452.1 | - | 1 - 2 | 248 | 87 | ||
FQ216455.1 | - | 1 - 2 | 252 | 87 | ||
FQ216484.1 | - | 1 - 2 | 230 | 87 | ||
FQ216496.1 | - | 1 - 2 | 251 | 87 | ||
FQ216525.1 | - | 1 - 2 | 251 | 87 | ||
FQ216543.1 | - | 1 - 2 | 248 | 87 | ||
FQ216631.1 | - | 1 - 2 | 251 | 87 | ||
FQ216704.1 | - | 1 - 2 | 251 | 87 | ||
FQ216737.1 | - | 1 - 2 | 334 | 87 | ||
FQ217303.1 | - | 1 - 2 | 230 | 87 | ||
FQ217394.1 | - | 1 - 2 | 181 | 87 | ||
FQ223632.1 | - | 1 - 2 | 251 | 87 | ||
FQ223827.1 | - | 1 - 2 | 251 | 87 | ||
FQ224171.1 | - | 1 - 2 | 248 | 87 | ||
FQ224212.1 | - | 1 - 2 | 231 | 87 | ||
FQ224277.1 | - | 1 - 2 | 251 | 87 | ||
FQ224519.1 | - | 1 - 2 | 248 | 87 | ||
FQ224726.1 | - | 1 - 2 | 181 | 87 | ||
FQ224931.1 | - | 1 - 2 | 248 | 87 | ||
FQ224934.1 | - | 1 - 2 | 246 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
