Assay Details
Gene Symbol: Myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077656.1 | NP_001071124.1 | 2 - 3 | 265 | 117 | |
GenBank mRNA | CB809837.1 | - | 2 - 3 | 338 | 117 | |
FQ214580.1 | - | 2 - 3 | 392 | 117 | ||
FQ214604.1 | - | 2 - 3 | 310 | 117 | ||
FQ214608.1 | - | 2 - 3 | 270 | 117 | ||
FQ214631.1 | - | 2 - 3 | 252 | 117 | ||
FQ214712.1 | - | 2 - 3 | 273 | 117 | ||
FQ214723.1 | - | 2 - 3 | 273 | 117 | ||
FQ214727.1 | - | 2 - 3 | 273 | 117 | ||
FQ214742.1 | - | 2 - 3 | 273 | 117 | ||
FQ214939.1 | - | 2 - 3 | 270 | 117 | ||
FQ215050.1 | - | 2 - 3 | 273 | 117 | ||
FQ215083.1 | - | 2 - 3 | 270 | 117 | ||
FQ215201.1 | - | 2 - 3 | 270 | 117 | ||
FQ215435.1 | - | 2 - 3 | 270 | 117 | ||
FQ215492.1 | - | 2 - 3 | 252 | 117 | ||
FQ215563.1 | - | 2 - 3 | 270 | 117 | ||
FQ215610.1 | - | 2 - 3 | 273 | 117 | ||
FQ215774.1 | - | 2 - 3 | 270 | 117 | ||
FQ215848.1 | - | 2 - 3 | 270 | 117 | ||
FQ215875.1 | - | 2 - 3 | 270 | 117 | ||
FQ215933.1 | - | 2 - 3 | 273 | 117 | ||
FQ216034.1 | - | 2 - 3 | 273 | 117 | ||
FQ216109.1 | - | 2 - 3 | 270 | 117 | ||
FQ216181.1 | - | 2 - 3 | 252 | 117 | ||
FQ216218.1 | - | 2 - 3 | 270 | 117 | ||
FQ216229.1 | - | 2 - 3 | 540 | 117 | ||
FQ216407.1 | - | 2 - 3 | 441 | 117 | ||
FQ216452.1 | - | 2 - 3 | 270 | 117 | ||
FQ216455.1 | - | 2 - 3 | 274 | 117 | ||
FQ216484.1 | - | 2 - 3 | 252 | 117 | ||
FQ216496.1 | - | 2 - 3 | 273 | 117 | ||
FQ216525.1 | - | 2 - 3 | 273 | 117 | ||
FQ216543.1 | - | 2 - 3 | 270 | 117 | ||
FQ216631.1 | - | 2 - 3 | 273 | 117 | ||
FQ216704.1 | - | 2 - 3 | 273 | 117 | ||
FQ216737.1 | - | 2 - 3 | 356 | 117 | ||
FQ217303.1 | - | 2 - 3 | 252 | 117 | ||
FQ217394.1 | - | 2 - 3 | 203 | 117 | ||
FQ223632.1 | - | 2 - 3 | 273 | 117 | ||
FQ223827.1 | - | 2 - 3 | 273 | 117 | ||
FQ224171.1 | - | 2 - 3 | 270 | 117 | ||
FQ224212.1 | - | 2 - 3 | 253 | 117 | ||
FQ224277.1 | - | 2 - 3 | 273 | 117 | ||
FQ224519.1 | - | 2 - 3 | 270 | 117 | ||
FQ224726.1 | - | 2 - 3 | 203 | 117 | ||
FQ224931.1 | - | 2 - 3 | 270 | 117 | ||
FQ224934.1 | - | 2 - 3 | 268 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
