Assay Details
Gene Symbol: Myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020104.2 | NP_064489.1 | 1 - 2 | 63 | 66 | |
GenBank mRNA | CR755322.1 | - | 1 - 2 | 68 | 66 | |
CR756356.1 | - | 1 - 2 | 63 | 66 | ||
FQ214656.1 | - | 1 - 2 | 81 | 66 | ||
FQ215340.1 | - | 1 - 2 | 81 | 66 | ||
FQ216308.1 | - | 1 - 2 | 81 | 66 | ||
FQ216738.1 | - | 1 - 2 | 81 | 66 | ||
FQ217074.1 | - | 1 - 2 | 80 | 66 | ||
FQ217103.1 | - | 1 - 2 | 81 | 66 | ||
FQ217218.1 | - | 1 - 2 | 81 | 66 | ||
FQ217243.1 | - | 1 - 2 | 80 | 66 | ||
FQ217323.1 | - | 1 - 2 | 81 | 66 | ||
FQ217387.1 | - | 1 - 2 | 81 | 66 | ||
FQ217427.1 | - | 1 - 2 | 86 | 66 | ||
FQ217481.1 | - | 1 - 2 | 81 | 66 | ||
FQ217524.1 | - | 1 - 2 | 81 | 66 | ||
FQ217753.1 | - | 1 - 2 | 86 | 66 | ||
FQ217756.1 | - | 1 - 2 | 81 | 66 | ||
FQ217889.1 | - | 1 - 2 | 81 | 66 | ||
FQ217979.1 | - | 1 - 2 | 81 | 66 | ||
FQ218029.1 | - | 1 - 2 | 81 | 66 | ||
FQ218049.1 | - | 1 - 2 | 81 | 66 | ||
FQ223687.1 | - | 1 - 2 | 81 | 66 | ||
FQ223779.1 | - | 1 - 2 | 81 | 66 | ||
FQ223949.1 | - | 1 - 2 | 79 | 66 | ||
FQ223984.1 | - | 1 - 2 | 81 | 66 | ||
FQ224047.1 | - | 1 - 2 | 81 | 66 | ||
FQ224050.1 | - | 1 - 2 | 81 | 66 | ||
FQ224087.1 | - | 1 - 2 | 81 | 66 | ||
FQ224155.1 | - | 1 - 2 | 86 | 66 | ||
FQ224172.1 | - | 1 - 2 | 81 | 66 | ||
FQ224321.1 | - | 1 - 2 | 85 | 66 | ||
FQ224386.1 | - | 1 - 2 | 86 | 66 | ||
FQ224411.1 | - | 1 - 2 | 81 | 66 | ||
FQ224452.1 | - | 1 - 2 | 156 | 66 | ||
FQ224663.1 | - | 1 - 2 | 81 | 66 | ||
FQ224796.1 | - | 1 - 2 | 81 | 66 | ||
FQ224799.1 | - | 1 - 2 | 86 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_020104.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
