Assay Details
Gene Symbol: Mzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108470.1 | NP_001101940.1 | 2 - 3 | 1057 | 74 | |
XM_006228116.2 | XP_006228178.1 | 3 - 4 | 944 | 74 | ||
XM_008758767.1 | XP_008756989.1 | 3 - 4 | 1205 | 74 | ||
XM_008758768.1 | XP_008756990.1 | 2 - 3 | 1312 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
