Assay Details
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773665.1 | XP_008771887.1 | 3 - 4 | 488 | 139 | |
XM_217630.8 | XP_217630.3 | 2 - 3 | 528 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
