Assay Details
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009629.1 | NP_001009629.1 | 2 - 3 | 284 | 141 | |
GenBank mRNA | BC088281.1 | - | 2 - 3 | 284 | 141 | |
FQ216316.1 | - | 2 - 3 | 273 | 141 | ||
FQ232102.1 | - | 2 - 3 | 256 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
