Assay Details
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009629.1 | NP_001009629.1 | 5 - 6 | 630 | 105 | |
XM_006248734.2 | XP_006248796.1 | 4 - 5 | 521 | 105 | ||
GenBank mRNA | BC088281.1 | - | 5 - 6 | 630 | 105 | |
FQ216316.1 | - | 5 - 6 | 619 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
