Assay Details
Gene Symbol: Cadm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047102.1 | NP_001040567.1 | 6 - 7 | 945 | 78 | |
GenBank mRNA | DQ272744.1 | - | 6 - 7 | 945 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P548857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123269 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P548857 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P548857 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P548857 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P119669 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P548857 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21291 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37809 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52216 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 282 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51733 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80942 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30375 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47515 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104062 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60745 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2816 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39469 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78233 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5837 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24928 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10752687 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752683 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752688 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752681 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752685 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752680 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752684 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10752686 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10752679 | - | - |
Affymetrix Rat Clariom S Assay | PSR1100006489.rn.1 | TC1100001081.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100006488.rn.1 | TC1100001081.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10752679 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21370263 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351824 | - | - |
