Assay Details
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106418.1 | NP_001099888.1 | 6 - 7 | 876 | 117 | |
XM_006257698.2 | XP_006257760.1 | 6 - 7 | 760 | 117 | ||
XM_006257699.2 | XP_006257761.1 | 6 - 7 | 734 | 117 | ||
XM_008773706.1 | XP_008771928.1 | 5 - 6 | 2672 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
