Assay Details
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044252.1 | NP_001037717.1 | 7 - 8 | 736 | 76 | |
XM_006232014.2 | XP_006232076.1 | 6 - 7 | 964 | 76 | ||
XM_006232015.2 | XP_006232077.1 | 6 - 7 | 963 | 76 | ||
XM_006232016.2 | XP_006232078.1 | 7 - 8 | 750 | 76 | ||
GenBank mRNA | BC105901.1 | - | 3 - 4 | 320 | 76 | |
DQ504300.1 | - | 7 - 8 | 736 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
