Assay Details
Gene Symbol: Msmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080886.1 | NP_543162.1 | 1 - 2 | 59 | 70 | |
GenBank mRNA | BC063155.1 | - | 1 - 2 | 46 | 70 | |
D50559.1 | - | 1 - 2 | 59 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC063155.1, D50559.1, NM_080886.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
