Assay Details
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012598.2 | NP_036730.1 | 9 - 10 | 1612 | 72 | |
GenBank mRNA | BC081836.1 | - | 9 - 10 | 1612 | 72 | |
L03294.1 | - | 1597 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid biosynthetic process
response to cold
response to xenobiotic stimulus
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
positive regulation of sequestering of triglyceride
lipid catabolic process
triglyceride biosynthetic process
triglyceride catabolic process
very-low-density lipoprotein particle remodeling
triglyceride homeostasis
lipoprotein lipase activity
triglyceride lipase activity
signaling receptor binding
heparin binding
triglyceride binding
apolipoprotein binding
fatty acid biosynthetic process
response to cold
response to xenobiotic stimulus
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
positive regulation of sequestering of triglyceride
lipid catabolic process
triglyceride biosynthetic process
triglyceride catabolic process
very-low-density lipoprotein particle remodeling
triglyceride homeostasis
lipoprotein lipase activity
triglyceride lipase activity
signaling receptor binding
heparin binding
triglyceride binding
apolipoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1016480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95611 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1016480 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1016480 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1016480 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P151230 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1016480 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700436175 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13840 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96304 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36902 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700436175 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10791256 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791255 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791252 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791253 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791257 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791254 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791251 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791258 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791261 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10791250 | - | - |
Rat Genome 230 2.0 Array | 1386965_at | - | - |
Rat Genome 230 2.0 Array | 1386965_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386965_at | - | - |
Affymetrix HT Rat Focus Array | 1386965_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600011248.rn.1 | TC1600001446.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600011250.rn.1 | TC1600001446.rn.1 | RTA TC |
Rat Expression Set 230 | 1386965_PM_at | - | - |
Rat Expression Set 230 | 1386965_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791250 | - | - |
Rat Genome U34 Set | L03294_at | - | - |
Rat Genome U34 Set | L03294_g_at | - | - |
Rat Genome U34 Set | rc_AI237731_s_at | - | - |
Rat Genome U34 Set | L03294_g_at | - | - |
Rat Genome U34 Set | L03294_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21726260 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357053 | - | - |
RatRef-12 Expression BeadChip | ILMN_51990 | - | - |
