Assay Details
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108848.1 | NP_001102318.1 | 2 - 3 | 323 | 104 | |
GenBank mRNA | AB212679.1 | - | 1 - 2 | 171 | 104 | |
BC168972.1 | - | 2 - 3 | 329 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
