Assay Details
Gene Symbol: Ip6k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053316.2 | NP_445768.1 | 2 - 3 | 378 | 75 | |
XM_006243839.1 | XP_006243901.1 | 2 - 3 | 580 | 75 | ||
GenBank mRNA | AB049151.1 | - | 2 - 3 | 429 | 75 | |
BC078702.1 | - | 2 - 3 | 378 | 75 | ||
FQ211545.1 | - | 2 - 3 | 526 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
