Assay Details
Gene Symbol: Ppil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017383.1 | NP_001017383.1 | 11 - 12 | 840 | 64 | |
XM_008768892.1 | XP_008767114.1 | 11 - 12 | 876 | 64 | ||
GenBank mRNA | BC085890.1 | - | 11 - 12 | 840 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
