Assay Details
Gene Symbol: Ehhadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133606.1 | NP_598290.1 | 1 - 2 | 94 | 82 | |
GenBank mRNA | BC089777.1 | - | 1 - 2 | 83 | 82 | |
K03249.1 | - | 1 - 2 | 94 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
