Assay Details
Gene Symbol: Klhl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181473.2 | NP_852138.2 | 5 - 6 | 1562 | 118 | |
GenBank mRNA | AB101615.1 | - | 4 - 5 | 1414 | 118 | |
BC092204.1 | - | 5 - 6 | 1562 | 118 | ||
FQ219171.1 | - | 6 - 7 | 1645 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
