Assay Details
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008335.1 | NP_001008336.1 | 6 - 7 | 649 | 88 | |
XM_006248534.2 | XP_006248596.1 | 6 - 7 | 665 | 88 | ||
XM_006248535.2 | XP_006248597.1 | 6 - 7 | 665 | 88 | ||
GenBank mRNA | BC085859.1 | - | 6 - 7 | 649 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
