Assay Details
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023989.1 | NP_076479.1 | 3 - 4 | 316 | 69 | |
XM_008768805.1 | XP_008767027.1 | 3 - 4 | 447 | 69 | ||
GenBank mRNA | AF260129.1 | - | 3 - 4 | 316 | 69 | |
FQ210639.1 | - | 3 - 4 | 411 | 69 | ||
FQ212134.1 | - | 3 - 4 | 411 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
