Assay Details
Gene Symbol: Ahsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012898.4 | NP_037030.1 | 839 | 57 | ||
GenBank mRNA | AY325170.1 | - | 1373 | 57 | ||
BC091118.1 | - | 817 | 57 | |||
FQ209390.1 | - | 836 | 57 | |||
FQ209445.1 | - | 839 | 57 | |||
FQ209502.1 | - | 838 | 57 | |||
FQ209536.1 | - | 838 | 57 | |||
FQ209588.1 | - | 838 | 57 | |||
FQ209604.1 | - | 838 | 57 | |||
FQ209612.1 | - | 838 | 57 | |||
FQ209654.1 | - | 838 | 57 | |||
FQ209655.1 | - | 835 | 57 | |||
FQ209665.1 | - | 838 | 57 | |||
FQ209720.1 | - | 838 | 57 | |||
FQ209767.1 | - | 846 | 57 | |||
FQ209920.1 | - | 838 | 57 | |||
FQ210030.1 | - | 833 | 57 | |||
FQ210033.1 | - | 840 | 57 | |||
FQ210120.1 | - | 838 | 57 | |||
FQ210139.1 | - | 838 | 57 | |||
FQ210141.1 | - | 838 | 57 | |||
FQ210159.1 | - | 838 | 57 | |||
FQ210164.1 | - | 836 | 57 | |||
FQ210170.1 | - | 837 | 57 | |||
FQ210239.1 | - | 838 | 57 | |||
FQ210242.1 | - | 836 | 57 | |||
FQ210249.1 | - | 838 | 57 | |||
FQ210382.1 | - | 838 | 57 | |||
FQ210405.1 | - | 838 | 57 | |||
FQ210504.1 | - | 838 | 57 | |||
FQ210525.1 | - | 58 | ||||
FQ210709.1 | - | 837 | 57 | |||
FQ210731.1 | - | 835 | 57 | |||
FQ210739.1 | - | 838 | 57 | |||
FQ210764.1 | - | 839 | 57 | |||
FQ210791.1 | - | 1 - 2 | 232 | 57 | ||
FQ210802.1 | - | 501 | 57 | |||
FQ210896.1 | - | 836 | 57 | |||
FQ210953.1 | - | 838 | 57 | |||
FQ211020.1 | - | 838 | 57 | |||
FQ211090.1 | - | 838 | 57 | |||
FQ211177.1 | - | 838 | 57 | |||
FQ211179.1 | - | 838 | 57 | |||
FQ211303.1 | - | 838 | 57 | |||
FQ211304.1 | - | 838 | 57 | |||
FQ211379.1 | - | 838 | 57 | |||
FQ211387.1 | - | 628 | 57 | |||
FQ217075.1 | - | 836 | 57 | |||
FQ218089.1 | - | 837 | 57 | |||
FQ218223.1 | - | 838 | 57 | |||
FQ218236.1 | - | 838 | 57 | |||
FQ218262.1 | - | 838 | 57 | |||
FQ218264.1 | - | 837 | 57 | |||
FQ218269.1 | - | 838 | 57 | |||
FQ218319.1 | - | 437 | 57 | |||
FQ218393.1 | - | 838 | 57 | |||
FQ218410.1 | - | 645 | 57 | |||
FQ218424.1 | - | 835 | 57 | |||
FQ218479.1 | - | 838 | 57 | |||
FQ218480.1 | - | 839 | 57 | |||
FQ218487.1 | - | 838 | 57 | |||
FQ218493.1 | - | 838 | 57 | |||
FQ218494.1 | - | 331 | 57 | |||
FQ218500.1 | - | 838 | 57 | |||
FQ218502.1 | - | 838 | 57 | |||
FQ218509.1 | - | 836 | 57 | |||
FQ218531.1 | - | 839 | 57 | |||
FQ218533.1 | - | 838 | 57 | |||
FQ218569.1 | - | 836 | 57 | |||
FQ218570.1 | - | 838 | 57 | |||
FQ218585.1 | - | 837 | 57 | |||
FQ218609.1 | - | 839 | 57 | |||
FQ218713.1 | - | 836 | 57 | |||
FQ218744.1 | - | 821 | 57 | |||
FQ218765.1 | - | 838 | 57 | |||
FQ218796.1 | - | 838 | 57 | |||
FQ218858.1 | - | 835 | 57 | |||
FQ218872.1 | - | 839 | 57 | |||
FQ218892.1 | - | 5 - 6 | 838 | 57 | ||
FQ218931.1 | - | 837 | 57 | |||
FQ218951.1 | - | 839 | 57 | |||
FQ218970.1 | - | 836 | 57 | |||
FQ218974.1 | - | 838 | 57 | |||
FQ218980.1 | - | 838 | 57 | |||
FQ218981.1 | - | 836 | 57 | |||
FQ219018.1 | - | 838 | 57 | |||
FQ219028.1 | - | 838 | 57 | |||
FQ219042.1 | - | 837 | 57 | |||
FQ219060.1 | - | 839 | 57 | |||
FQ219086.1 | - | 839 | 57 | |||
FQ219099.1 | - | 838 | 57 | |||
FQ219110.1 | - | 837 | 57 | |||
FQ219130.1 | - | 834 | 57 | |||
FQ219141.1 | - | 839 | 57 | |||
FQ219164.1 | - | 840 | 57 | |||
FQ219203.1 | - | 838 | 57 | |||
FQ219235.1 | - | 838 | 57 | |||
FQ219239.1 | - | 839 | 57 | |||
FQ219274.1 | - | 837 | 57 | |||
FQ219295.1 | - | 838 | 57 | |||
FQ219300.1 | - | 839 | 57 | |||
FQ219311.1 | - | 838 | 57 | |||
FQ219320.1 | - | 839 | 57 | |||
FQ219487.1 | - | 58 | ||||
FQ219488.1 | - | 838 | 57 | |||
FQ219536.1 | - | 837 | 57 | |||
FQ219551.1 | - | 838 | 57 | |||
FQ219606.1 | - | 839 | 57 | |||
FQ219608.1 | - | 838 | 57 | |||
FQ219616.1 | - | 573 | 57 | |||
FQ219617.1 | - | 839 | 57 | |||
FQ219643.1 | - | 838 | 57 | |||
FQ219660.1 | - | 837 | 57 | |||
FQ219663.1 | - | 838 | 57 | |||
FQ219668.1 | - | 840 | 57 | |||
FQ219672.1 | - | 838 | 57 | |||
FQ219726.1 | - | 839 | 58 | |||
FQ219772.1 | - | 838 | 57 | |||
FQ219774.1 | - | 838 | 57 | |||
M29758.1 | - | 839 | 57 | |||
X63446.1 | - | 797 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
