Assay Details
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053842.1 | NP_446294.1 | 2 - 3 | 454 | 65 | |
XM_006248658.2 | XP_006248720.1 | 2 - 3 | 491 | 65 | ||
XM_006248659.2 | XP_006248721.1 | 2 - 3 | 491 | 65 | ||
XM_008768848.1 | XP_008767070.1 | 2 - 3 | 491 | 65 | ||
GenBank mRNA | FQ211642.1 | - | 2 - 3 | 494 | 65 | |
M64300.1 | - | 2 - 3 | 454 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
