Assay Details
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134789.1 | NP_001128261.1 | 3 - 4 | 343 | 88 | |
GenBank mRNA | BQ210721.1 | - | 2 - 3 | 88 | ||
CB758009.1 | - | 3 - 4 | 357 | 88 | ||
CF113682.1 | - | 3 - 4 | 316 | 88 | ||
FQ222889.1 | - | 3 - 4 | 341 | 88 |
Gene Symbol: LOC100912534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751083.2 | XP_003751131.1 | 3 - 4 | 357 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
