Assay Details
Gene Symbol: Gnb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248650.2 | XP_006248712.1 | 1 - 2 | 178 | 53 | |
XM_006248654.2 | XP_006248716.1 | 1 - 2 | 179 | 53 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248650.2, XM_006248654.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
