Assay Details
Gene Symbol: Nrep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254595.1 | XP_006254657.1 | 4 | 171 | ||
GenBank mRNA | AY724475.1 | - | 4 | 1421 | 153 | |
FQ213250.1 | - | 1 | 171 | |||
FQ214488.1 | - | 4 | 171 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
