Assay Details
Gene Symbol: Rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014149.1 | NP_001014171.1 | 1 - 2 | 335 | 59 | |
GenBank mRNA | BC089946.1 | - | 1 - 2 | 335 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC089946.1, NM_001014149.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
