Assay Details
Gene Symbol: Rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014149.1 | NP_001014171.1 | 2 - 3 | 577 | 85 | |
GenBank mRNA | BC089946.1 | - | 2 - 3 | 577 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
