Assay Details
Gene Symbol: Slc39a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024745.1 | NP_001019916.1 | 6 - 7 | 1590 | 62 | |
GenBank mRNA | BC097493.1 | - | 6 - 7 | 1590 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
