Assay Details
Gene Symbol: Fgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254651.2 | XP_006254713.1 | 1 - 2 | 1348 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006254651.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
