Assay Details
Gene Symbol: Fgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254650.2 | XP_006254712.1 | 2 - 3 | 534 | 75 | |
XM_006254654.2 | XP_006254716.1 | 2 - 3 | 538 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006254650.2, XM_006254654.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
