Assay Details
Gene Symbol: Tmco6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106154.1 | NP_001099624.1 | 3 - 4 | 449 | 55 | |
GenBank mRNA | BC166835.1 | - | 3 - 4 | 311 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
